S-Carboxymethyl-L-cysteine, 98%
S-Carboxymethyl-L-cysteine, 98%
S-Carboxymethyl-L-cysteine, 98%
Thermo Scientific Chemicals

S-Carboxymethyl-L-cysteine, 98%

CAS: 638-23-3 | C5H9NO4S | 179.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 225810250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS638-23-3
IUPAC Name(2R)-2-amino-3-[(carboxymethyl)sulfanyl]propanoic acid
Molecular FormulaC5H9NO4S
InChI KeyGBFLZEXEOZUWRN-VKHMYHEASA-N
SMILESN[C@@H](CSCC(O)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance of solution(1% soln. in 0.1 N HCl, gently warmed)
Appearance of solutionClear colorless
Residue after ignition=<0.2 %
Heavy metals (as Pb)=<10 ppm
Appearance (Color)White to almost white
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only