N-Acetyl-D-galactosamine, 97%, Thermo Scientific Chemicals
N-Acetyl-D-galactosamine, 97%, Thermo Scientific Chemicals
N-Acetyl-D-galactosamine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Acetyl-D-galactosamine, 97%, Thermo Scientific Chemicals

Quantity:
500 mg
1 g
Packaging:
Glass bottle
Catalog number 225850010
Price (JPY)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS14215-68-0
IUPAC NameN-[(3R,4R,5R,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide
Molecular FormulaC8H15NO6
InChI KeyOVRNDRQMDRJTHS-KEWYIRBNSA-N
SMILESCC(=O)N[C@H]1C(O)O[C@H](CO)[C@H](O)[C@@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Specific optical rotationafter 5 h, on dry substance
Solubility(10 % in water) Clear colorless
Infrared spectrumConforms
Appearance (Color)White to off-white
Appearance (Form)Crystalline powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only