L(-)-Arabitol, 99%
L(-)-Arabitol, 99%
L(-)-Arabitol, 99%
Thermo Scientific Chemicals

L(-)-Arabitol, 99%

CAS: 7643-75-6 | C5H12O5 | 152.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 225860250
Price (EUR)
165,00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
165,00
Each
Chemical Identifiers
CAS7643-75-6
IUPAC Namepentane-1,2,3,4,5-pentol
Molecular FormulaC5H12O5
InChI KeyHEBKCHPVOIAQTA-UHFFFAOYNA-N
SMILESOCC(O)C(O)C(O)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point99°C to 104°C
HPLC>=98.5 %
Specific optical rotation-10° to -14° (20°C, 589 nm) (c=5,8% borax)
Appearance (Color)White to almost white
Appearance (Form)Powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only