Maltotriose, 93%
Maltotriose, 93%
Maltotriose, 93%
Thermo Scientific Chemicals

Maltotriose, 93%

CAS: 1109-28-0 | C18H32O16 | 504.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
25 g
Catalog number 225940250
Price (USD)
591.65
Special offer
Online exclusive
Ends: 28-Dec-2026
696.00
Save 104.35
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
591.65
Special offer
Online exclusive
Ends: 28-Dec-2026
696.00
Save 104.35
Each
Chemical Identifiers
CAS1109-28-0
IUPAC Name2-{[4,5-dihydroxy-2-(hydroxymethyl)-6-{[4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxan-3-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular FormulaC18H32O16
InChI KeyFYGDTMLNYKFZSV-UHFFFAOYNA-N
SMILESOCC1OC(OC2C(O)C(O)C(OC3C(O)C(O)C(O)OC3CO)OC2CO)C(O)C(O)C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Specific optical rotation+155° to +165° (20°C, 589 nm) (c=1, water)
Infrared spectrumConforms
HPLC>=91 %
Water=<6 % (K.F.)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only