L-Cysteic acid, 99%
L-Cysteic acid, 99%
L-Cysteic acid, 99%
Thermo Scientific Chemicals

L-Cysteic acid, 99%

CAS: 498-40-8 | C3H7NO5S | 169.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 226120100
Price (USD)
90.10
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
90.10
Each
Chemical Identifiers
CAS498-40-8
IUPAC Name2-amino-3-sulfopropanoic acid
Molecular FormulaC3H7NO5S
InChI KeyXVOYSCVBGLVSOL-UHFFFAOYNA-N
SMILESNC(CS(O)(=O)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Specific optical rotation+7° to +9° (20°C, 589 nm) (c=5, H2O)
Infrared spectrumConforms
Water=<1 % (K.F.)
Solubility(5 % in water) Clear colorless
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only