N6-Benzyladenosine
N6-Benzyladenosine
N6-Benzyladenosine
Thermo Scientific Chemicals

N6-Benzyladenosine

Catalog NumberQuantity
2264200101 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 226420010
Price (JPY)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS4294-16-0
IUPAC Name(2R,3R,4S,5R)-2-[6-(benzylamino)-9H-purin-9-yl]-5-(hydroxymethyl)oxolane-3,4-diol
Molecular FormulaC17H19N5O4
InChI KeyMRPKNNSABYPGBF-ABJKBEJCNA-N
SMILESOC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N1C=NC2=C(NCC3=CC=CC=C3)N=CN=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
HPLC>=97.5 %
Appearance (Color)White
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only