Thermo Scientific™

2'-Deoxyguanosine hydrate, 99+%

100mg, Each
Thermo Scientific™

2'-Deoxyguanosine hydrate, 99+%

100mg, Each
Quantity

Chemical Identifiers

CAS
207121-55-9
HPLC
>=99.0 %
UV spectrophotometry
molar absorptivity 13700 +/- 4%, on dry basis
Appearance (Form)
Crystalline powder
Water
=<8.0 % (K.F.)
Appearance (Color)
White to off-white

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
RUO – Research Use Only

Figures