1-Hexadecylpyridinium chloride monohydrate, 96.0-101.0%
1-Hexadecylpyridinium chloride monohydrate, 96.0-101.0%
1-Hexadecylpyridinium chloride monohydrate, 96.0-101.0%
Thermo Scientific Chemicals

1-Hexadecylpyridinium chloride monohydrate, 96.0-101.0%

CAS: 6004-24-6 | C21H40ClNO | 358.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 226991000
Price (USD)
53.65
Special offer
Online exclusive
Ends: 28-Dec-2026
63.50
Save 9.85
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
53.65
Special offer
Online exclusive
Ends: 28-Dec-2026
63.50
Save 9.85
Each
Chemical Identifiers
CAS6004-24-6
IUPAC Name1-hexadecyl-1,4-dihydropyridin-4-ylium hydrate chloride
Molecular FormulaC21H40ClNO
InChI KeyNFCRBQADEGXVDL-UHFFFAOYSA-M
SMILESO.[Cl-].CCCCCCCCCCCCCCCCN1C=C[CH+]C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White to almost white
Appearance (Form)Powder
Titration96.0 to 101.0 %
Water4.5 to 5.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only