D-Serine, 98%
D-Serine, 98%
D-Serine, 98%
Thermo Scientific Chemicals

D-Serine, 98%

CAS: 312-84-5 | C3H7NO3 | 105.093 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
Catalog number 227070250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS312-84-5
IUPAC Name2-amino-3-hydroxypropanoic acid
Molecular FormulaC3H7NO3
InChI KeyMTCFGRXMJLQNBG-UHFFFAOYNA-N
SMILESNC(CO)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Sulfated ash=<0.1 %
Specific optical rotation-13.0° to -16.0° (20°C, 589 nm) (c=10, 2 N HCl)
Infrared spectrumConforms
Appearance (Color)White to off-white
Loss on drying=<0.3 % (105°C, 3 h)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only