Colchicine, 97%
Colchicine, 97%
Colchicine, 97%
Thermo Scientific Chemicals

Colchicine, 97%

Colchicine, 7alphah-colchicine, CAS # 64-86-8, is a laboratory grade formulation of the anti-inflammatory plant alkaloid drug. | CAS: 64-86-8 | C22H25NO6 | 399.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
1 g
5 g
25 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 227120010
Price (EUR)
126,65
Online exclusive
141,00
Save 14,35
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
126,65
Online exclusive
141,00
Save 14,35
Each
Chemical Identifiers
CAS64-86-8
IUPAC NameN-[(10S)-3,4,5,14-tetramethoxy-13-oxotricyclo[9.5.0.0²,⁷]hexadeca-1(16),2,4,6,11,14-hexaen-10-yl]acetamide
Molecular FormulaC22H25NO6
InChI KeyIAKHMKGGTNLKSZ-INIZCTEOSA-N
SMILESCOC1=C(OC)C(OC)=C2C(CC[C@H](NC(C)=O)C3=CC(=O)C(OC)=CC=C23)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Specific optical rotationon anhydrous and solvent-free substance
Infrared spectrumConforms
HPLC>=96.0 % on anhydrous and solvent-free substance
Specific optical rotation-240° to -250° (20°C, 589 nm) (c=1, alcohol)
Appearance (Color)Light yellow
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

  • Colchicine (7alphah-colchicine) is an alkaloid found in Colchicum autumnale that has been shown to have anti-gout and anti-inflammatory properties
  • Colchicine binds tubulin and prevents the migration of immune cells, reducing inflammation

Applications

  • Colchicine is an anti-mitotic; may be used in the study of cell proliferation and migration
  • Intended for in vivo and in vitro research applications
  • For research use only, not intended to treat disease
RUO – Research Use Only