D-Valine, 98+%
D-Valine, 98+%
D-Valine, 98+%
Thermo Scientific Chemicals

D-Valine, 98+%

CAS: 640-68-6 | C5H11NO2 | 117.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass Bottle
Catalog number 227220050
Price (USD)
32.65
Special offer
Online exclusive
Ends: 28-Dec-2026
38.30
Save 5.65
Each
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
32.65
Special offer
Online exclusive
Ends: 28-Dec-2026
38.30
Save 5.65
Each
Chemical Identifiers
CAS640-68-6
IUPAC Name(2R)-2-amino-3-methylbutanoic acid
Molecular FormulaC5H11NO2
InChI KeyKZSNJWFQEVHDMF-SCSAIBSYSA-N
SMILESCC(C)[C@@H](N)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.5 %
Sulfated ash=<0.2 %
Appearance (Form)Crystalline powder
Titration with HClO498.0 to 102.0 %
Loss on drying=<0.5 % (105°C, 4 hrs)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only