Thermo Scientific Chemicals

D-Valine, +98%

Catalog number: 227220250
25 g, Glass Bottle, Each
Thermo Scientific Chemicals

D-Valine, +98%

Catalog number: 227220250
25 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 227220250
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
640-68-6
IUPAC Name
(2R)-2-amino-3-methylbutanoic acid
Molecular Formula
C5H11NO2
InChI Key
KZSNJWFQEVHDMF-SCSAIBSYSA-N
SMILES
CC(C)[C@@H](N)C(O)=O
Appearance (Color)
White to off-white
Infrared spectrum
Conforms
Water
=<0.5 %
Sulfated ash
=<0.2 %
Appearance (Form)
Crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text