D(-)-Isoascorbic acid, 98%
D(-)-Isoascorbic acid, 98%
D(-)-Isoascorbic acid, 98%
Thermo Scientific Chemicals

D(-)-Isoascorbic acid, 98%

CAS: 89-65-6 | C6H7NaO6 | 198.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
Catalog number 227270025
Price (USD)
135.65
Special offer
Online exclusive
Ends: 28-Dec-2026
159.00
Save 23.35
Each
Quantity:
2.5 kg
Request bulk or custom format
Price (USD)
135.65
Special offer
Online exclusive
Ends: 28-Dec-2026
159.00
Save 23.35
Each
Chemical Identifiers
CAS89-65-6
IUPAC Namesodium 5-[(1S)-1,2-dihydroxyethyl]-3-hydroxy-2,4-dioxooxolan-3-ide
Molecular FormulaC6H7NaO6
InChI KeyIFVCRSPJFHGFCG-HXPAKLQESA-N
SMILES[Na+].OC[C@H](O)C1OC(=O)[C-](O)C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Melting point164°C to 172°C
Lead (Pb)=<10 ppm
Appearance (Color)White to light yellow
Loss on drying=<0.4 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only