(-)-Erythromycin, 98%
(-)-Erythromycin, 98%
(-)-Erythromycin, 98%
Thermo Scientific Chemicals

(-)-Erythromycin, 98%

Erythromycin (E-Mycin or erythrocin), CAS # 114-07-8, is a broad-spectrum antibiotic.
Have Questions?
Catalog NumberQuantityPackaging
22733025025 gPlastic Bottle
Catalog number 227330250
Price (USD)
191.00
Each
Quantity:
25 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (USD)
191.00
Each
Chemical Identifiers
CAS114-07-8
IUPAC Name(3R,4S,5S,6R,7R,9R,11R,12R,13S,14R)-6-{[(2S,3R,4S,6R)-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy}-14-ethyl-7,12,13-trihydroxy-4-{[(2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyloxan-2-yl]oxy}-3,5,7,9,11,13-hexamethyl-1-oxacyclotetradecane-2,10-dione
Molecular FormulaC37H67NO13
InChI KeyULGZDMOVFRHVEP-RWJQBGPGSA-N
SMILESCC[C@H]1OC(=O)[C@H](C)[C@@H](O[C@H]2C[C@@](C)(OC)[C@@H](O)[C@H](C)O2)[C@H](C)[C@@H](O[C@@H]2O[C@H](C)C[C@@H]([C@H]2O)N(C)C)[C@](C)(O)C[C@@H](C)C(=O)[C@H](C)[C@@H](O)[C@]1(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Specific optical rotation-71° to -78° (20°C, 589 nm) (c=2, C2H5OH, 30 minutes) on dry substance
Appearance (Color)White to light yellow
Infrared spectrumConforms
Water=<6.5 % (K.F.)
Appearance (Form)Powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Erythromycin is a macrolide antibiotic that works by binding the 50S ribosomal subunit and inhibiting polypeptide formation

• Erythromycin was isolated from Saccharopolyspora erythraea

Applications

• Erythromycin can be used to disrupt the growth of gram-positive and gram-negative bacteria

• Erythromycin has been shown to have motor effects in the gastrointestinal system and can be used for laboratory research on migrating motor complex activity

RUO – Research Use Only

General References:

  1. Dreno, B.; Martin, R.; Moyal, D.; Henley, J.B.; Khammari, A.; Seité, S. Skin microbiome and acne vulgaris: Staphylococcus, a new actor in acne. Exp Dermatol. 2017, 26(9), 798-803.
  2. Wu, F.; Tokach, M.D.; DeRouchey, J.M.; Dritz, S.S.; Woodworth, J.C.; Goodband, R.D.; Chitakasempornkul, K.; Bello, N.M.; Capps, K.; Remfry, S.; Scott, H.M.; Nagaraja, T.G.; Apley, M.D.; Amachawadi, R.G. Effects of Tylosin Administration Routes on the Prevalence of Antimicrobial Resistance Among Fecal Enterococci of Finishing Swine. Foodborne Pathog Dis. 2019, 16(5), 309-316.