Polyvinylpyrrolidone, average M.W. 8.000, K16-18
Polyvinylpyrrolidone, average M.W. 8.000, K16-18
Polyvinylpyrrolidone, average M.W. 8.000, K16-18
Thermo Scientific Chemicals

Polyvinylpyrrolidone, average M.W. 8.000, K16-18

CAS: 9003-39-8 | (C6H9NO)n | 111.14 g/mol
数量:
500 g
2.5 kg
パッケージ:
Plastic bottle
製品番号(カタログ番号) 227460025
価格(JPY)
-
数量:
2.5 kg
パッケージ:
Plastic bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS9003-39-8
Molecular Formula(C6H9NO)n
SMILES*-CC(-*)N1CCCC1=O
MDL NumberMFCD01076626
CAS9003-39-8
さらに表示
Appearance of solution(Ph. Eur.) Clear and not more intensely colored than ref. B6
Appearance of solution(Ph. Eur.) Clear and not more intensely colored than ref. BY6
Appearance of solution(Ph. Eur.) Clear and not more intensely colored than ref. R6
K-value16 to 18 (5 % solids w/v aq. soln.)
Appearance of solution(Ph. Eur.) Clear and not more intensely colored than ref. B6
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only