Auramine O, pure, biological stain
Auramine O, pure, biological stain
Auramine O, pure, biological stain
Thermo Scientific Chemicals

Auramine O, pure, biological stain

CAS: 2465-27-2 | C17H22ClN3 | 303.834 g/mol
Have Questions?
Catalog NumberQuantity
22752050050 g
Catalog number 227520500
Price (USD)
121.65
Online exclusive
135.00
Save 13.35
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
121.65
Online exclusive
135.00
Save 13.35
Each
Chemical Identifiers
CAS2465-27-2
IUPAC Namehydrogen 4-[4-(dimethylamino)benzenecarboximidoyl]-N,N-dimethylaniline chloride
Molecular FormulaC17H22ClN3
InChI KeyKSCQDDRPFHTIRL-UHFFFAOYSA-N
SMILES[H+].[Cl-].CN(C)C1=CC=C(C=C1)C(=N)C1=CC=C(C=C1)N(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Dye content>=60 %
Appearance (Form)Powder
Appearance (Color)Yellow to ochre
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only