Cinchonine, 98+%
Cinchonine, 98+%
Cinchonine, 98+%
Thermo Scientific Chemicals

Cinchonine, 98+%

CAS: 118-10-5 | C19H22N2O | 294.40 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 227810250
Price (EUR)
43,65
Online exclusive
48,70
Save 5,05
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
43,65
Online exclusive
48,70
Save 5,05
Each
Chemical Identifiers
CAS118-10-5
IUPAC Name(S)-[(2R,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl](quinolin-4-yl)methanol
Molecular FormulaC19H22N2O
InChI KeyKMPWYEUPVWOPIM-FRYPIZGFNA-N
SMILES[H][C@@]1(CN2CCC1C[C@]2([H])[C@@H](O)C1=CC=NC2=CC=CC=C12)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white to light beige
Appearance (Form)Crystalline powder or needles
Infrared spectrumConforms
Melting point255°C to 265°C
Titration with HClO498.0 to 102.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only