D(-)-Glutamic acid, 99+%
D(-)-Glutamic acid, 99+%
D(-)-Glutamic acid, 99+%
Thermo Scientific Chemicals

D(-)-Glutamic acid, 99+%

CAS: 6893-26-1 | C5H9NO4 | 147.13 g/mol
Have Questions?
Catalog NumberQuantityPackaging
22789025025 gGlass bottle
Catalog number 227890250
Price (USD)
132.65
Special offer
Online exclusive
Ends: 30-Jun-2026
156.00
Save 23.35 (15%)
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
132.65
Special offer
Online exclusive
Ends: 30-Jun-2026
156.00
Save 23.35 (15%)
Each
Chemical Identifiers
CAS6893-26-1
IUPAC Name2-aminopentanedioic acid
Molecular FormulaC5H9NO4
InChI KeyWHUUTDBJXJRKMK-UHFFFAOYNA-N
SMILESNC(CCC(O)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Infrared spectrumConforms
Sulfated ash=<0.1 %
Other amino acids=<0.5 % (TLC)
Titration with NaOH99.0 to 101.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only