ART™ Barrier Specialty Pipette tips
ART™ Barrier Specialty Pipette tips
ART™ Barrier Specialty Pipette tips
Thermo Scientific™

ART™ Barrier Specialty Pipette tips

Perform pipetting accurately and consistently with these barrier extended length pipette tips, which seal when exposed to potential contaminants.

Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)ModelTip StyleFit for PurposeUnit Size
22791000 μLART 1000ErgonomicSterile and PCR ReadyCase of 3200
23801000 μLART 1000 ERGO-FErgonomicSterile and PCR ReadyEach
2770200 μLART 200UErgonomicSterile and PCR ReadyCase of 4800
2770PK
also known as 2770
200 μLART 200UErgonomicSterile and PCR ReadyPack of 960
278050 μLART 50UErgonomicSterile and PCR ReadyCase of 4800
2780PK
also known as 2780
50 μLART 50UErgonomicSterile and PCR ReadyPack of 960
2279PK
also known as 2279
1000 μLART 1000ErgonomicSterile and PCR ReadyPack of 800
2140PK
also known as 2140
10 μLART 10 REACHExtended LengthSterile and PCR ReadyPack of 960
21490.5 to 20 μLART 20EMicro PointSterile and PCR ReadyCase of 4800
2149PK
also known as 2149
0.5 to 20 μLART 20EMicro PointSterile and PCR ReadyPack of 960
Catalog number 2279
Price (USD)
506.80
Online exclusive
724.00
Save 217.20
Case of 3200
Volume (Metric):
1000 μL
Model:
ART 1000
Tip Style:
Ergonomic
Fit for Purpose:
Sterile and PCR Ready
Unit Size:
Case of 3200
Price (USD)
506.80
Online exclusive
724.00
Save 217.20
Case of 3200
Perform applications consistently and accurately by using Thermo Scientific™ ART™ Barrier Specialty Pipette Tips, which seal when exposed to potential contaminants, trapping them inside the barrier. ART barrier specialty tips are designed to aid in pipetting for a variety of applications. Extended length, Thermo Scientific™ REACH™ tips provide an extra level of contamination control.

Features:

  • Sterile tips are DNase, RNase, DNA, ATP, PCR Inhibitor, and Endotoxin Free
  • Available in sterile, nonsterile and in various packaging options
  • Manufactured according to the industry's highest quality standard

Designed to Aid in Pipetting for a Variety of Applications:

Extended length: to prevent contamination by keeping the pipette shaft from entering the sample vessel

Gel loading: for loading acrylamide or agarose gels

Wide orifice genomic: for genomic DNA, fragile cell lines and other viscous materials

Solvent safe: for acids, bases and aggressive organic solvents

Compatible with:

SoftFit™ pipette tips provide precise and consistent pipetting with multichannel pipettes

Ergonomic pipette tips are designed for Gilson Pipetman™ pipettes and Thermo Scientific™ Finnpipette™ Focus pipettes

Specifications
Clean ClaimsDNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free
DescriptionART 1000, Filtered, Sterile, Lift-off Lid Rack
Length (English)3.13 in.
Length (Metric)7.9cm
ModelART 1000
FilteredFiltered
Fit for PurposeSterile and PCR Ready
For Use With (Equipment)Gilson Pipetman, Thermo Scientific Finnpipette Focus
FormatLift-off Lid Rack
Product LineART
SterilitySterile
Tip StyleErgonomic
Volume (Metric)1000 μL
Unit SizeCase of 3200