ADA, 99%, for biochemistry
ADA, 99%, for biochemistry
ADA, 99%, for biochemistry
Thermo Scientific Chemicals

ADA, 99%, for biochemistry

CAS: 26239-55-4 | C6H10N2O5 | 190.155 g/mol
数量:
25 g
100 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 228010250
価格(JPY)
-
数量:
25 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS26239-55-4
IUPAC Name2-[(carbamoylmethyl)(carboxylatomethyl)azaniumyl]acetate
Molecular FormulaC6H9N2O5
InChI KeyQZTKDVCDBIDYMD-UHFFFAOYSA-M
SMILESNC(=O)C[NH+](CC([O-])=O)CC([O-])=O
さらに表示
Heavy metals (as Pb)=<5 ppm
Sulfated ash=<0.1 %
Loss on drying=<0.5 % (110°C, 5 hrs)
UV(9 % in 1 M NaOH)
Appearance (Form)Crystalline powder
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only