HyPURITY™ Cyano HPLC Columns
HyPURITY™ Cyano HPLC Columns
HyPURITY HPLC Columns are highly characterized, tested, and validated.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)
22805-254630250 mm
22805-154630150 mm
Catalog number 22805-254630
Price (USD)
-
Length (Metric):
250 mm
Thermo Scientific™ HyPURITY™ Cyano HPLC Columns are based on the HyPURITY 190Å silica—the highest quality silica available and essentially metal-free with exceptionally stablity at high and low pH. The rugged and robust manufacturing process ensures reproducibility and optimum quality, batch after batch. Within the manufacturing process, extra care is taken to ensure an extensively homogenous silica surface. This surface is a key precursor in the preparation of uniform bonded phases. Cyano functionality is generally less hydrophobic and therefore compounds are less retained compared to other alkyl phases, though it does have strong dipole interaction. Additionally, cyano can be operated under either reversed-phase or normal phase / HILIC conditions.
Specifications
Column FormatAnalytical Column
Column TypeMixed Mode
EndcappedYes
For Use WithSVL™ Stirrer Bearing
Packing MaterialSilica
Product TypeTorion Body
Stationary PhaseCyano (CN)
Surface Area200 m²/g
USP TypeL10
Carbon Load4%
Diameter (Metric)4.6 mm
Length (Metric)250 mm
MaterialGlass
Particle Size5 μm
PhaseReversed Phase, Normal Phase, Polar Selectivity
Pore Size190 Å
Unit SizeEach