Carbonyl cyanide 3-chlorophenylhydrazone, 98%
Carbonyl cyanide 3-chlorophenylhydrazone, 98%
Carbonyl cyanide 3-chlorophenylhydrazone, 98%
Thermo Scientific Chemicals

Carbonyl cyanide 3-chlorophenylhydrazone, 98%

CAS: 555-60-2 | C9H5ClN4 | 204.62 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
1 g
Packaging:
Glass bottle
Catalog number 228131000
Price (USD)
37.00
Each
Quantity:
100 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
37.00
Each
Chemical Identifiers
CAS555-60-2
IUPAC NameN-(3-chlorophenyl)-1-cyanomethanecarbohydrazonoyl cyanide
Molecular FormulaC9H5ClN4
InChI KeyUGTJLJZQQFGTJD-UHFFFAOYSA-N
SMILESClC1=CC=CC(NN=C(C#N)C#N)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=97.5 %
Appearance (Color)Yellow to orange to orange-brown
Infrared spectrumConforms
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only