I used the Thermo Scientific Power Stainer for staining of my gel. I see non-specific spots or splotches of stain in the polyacrylamide background not associated with protein. Can you help me troubleshoot?
Here are possible causes and solutions:
- Inefficient washing of gel. Wash Mini gel at least once for 5 mins and Midi gel twice for 5 mins each in water. When staining 2 Mini gels simultaneously, wash each Mini gel 2 x 5 mins in water.
- Insufficient staining / destaining time. Add additional 30 seconds to 1 minute of staining / destaining time. Or, destain the gel in water for additional time.
- Air bubbles trapped between gel and the pads. When assembling staining stack, use a roller or pipette to remove any air bubbles between the gel and the pads.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
- Inefficient washing of gel. Wash Mini gel at least once for 5 mins and Midi gel twice for 5 mins each in water. When staining 2 Mini gels simultaneously, wash each Mini gel 2 x 5 mins in water.
- Insufficient staining / destaining time. Add additional 30 seconds to 1 minute of staining / destaining time. Or, destain the gel in water for additional time.
- Air bubbles trapped between gel and the pads. When assembling staining stack, use a roller or pipette to remove any air bubbles between the gel and the pads.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.