Brij™ L23 PHARMA-PA-(MV), previous name Brij™ 35
Brij™ L23 PHARMA-PA-(MV), previous name Brij™ 35
Brij™ L23 PHARMA-PA-(MV), previous name Brij™ 35
Thermo Scientific Chemicals

Brij™ L23 PHARMA-PA-(MV), previous name Brij™ 35

CAS: 9002-92-0 | C14H30O2 | 230.39 g/mol
製品番号(カタログ番号)数量パッケージ
2283400515 kgPlastic drum
製品番号(カタログ番号) 228340051
価格(JPY)
-
数量:
5 kg
パッケージ:
Plastic drum
一括またはカスタム形式をリクエストする
化学物質識別子
CAS9002-92-0
MDL NumberMFCD00043063
Infrared spectrumConforms
Hydroxyl value40 to 60 mg KOH/g
Acid value=<5 mg KOH/g
Impurity=<10 ppm free Dioxane
Appearance (Color)White
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only