Brij™ L23 PHARMA-PA-(MV), previous name Brij™ 35
Brij™ L23 PHARMA-PA-(MV), previous name Brij™ 35
Brij™ L23 PHARMA-PA-(MV), previous name Brij™ 35
Thermo Scientific Chemicals

Brij™ L23 PHARMA-PA-(MV), previous name Brij™ 35

CAS: 9002-92-0 | C14H30O2 | 230.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
500 g
1 kg
5 kg
Packaging:
View all
HDPE plastic bottle
Catalog number 228342500
Price (USD)
50.70
Each
Quantity:
250 g
Packaging:
HDPE plastic bottle
Request bulk or custom format
Price (USD)
50.70
Each
Chemical Identifiers
CAS9002-92-0
MDL NumberMFCD00043063
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Hydroxyl value40 to 60 mg KOH/g
Acid value=<5 mg KOH/g
Impurity=<10 ppm free Dioxane
Appearance (Color)White
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only