Eriochrome™ Black T, pure, indicator grade
Eriochrome™ Black T, pure, indicator grade
Eriochrome™ Black T, pure, indicator grade
Thermo Scientific Chemicals

Eriochrome™ Black T, pure, indicator grade

CAS: 1787-61-7 | C20H12N3NaO7S | 461.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
1 kg
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 228361000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1787-61-7
IUPAC Namesodium (4Z)-4-[2-(1-hydroxynaphthalen-2-yl)hydrazin-1-ylidene]-7-nitro-3-oxo-3,4-dihydronaphthalene-1-sulfonate
Molecular FormulaC20H12N3NaO7S
InChI KeyJHUJLRKQZAPSDP-GXTSIBQPSA-M
SMILES[Na+].OC1=C2C=CC=CC2=CC=C1N\N=C1/C(=O)C=C(C2=CC(=CC=C12)[N+]([O-])=O)S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
UV spectrumComplies
Suitability(for compl. det. of metals) Passes test
Appearance (Form)Powder
Infrared spectrumConforms
Appearance (Color)Brown to black
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only