Pierce™ Mini Gel Power Staining Kit
Thermo Scientific™

Pierce™ Mini Gel Power Staining Kit

The Thermo Scientific™ Pierce Power Staining kits are designed for fast Coomassie dye staining of protein gels. The Pierce Mini Gel Power Staining Kit enables rapid Coomassie dye staining of proteins mini-size polyacrylamide gels and the removal of unbound stain from the gel matrix.
Have Questions?
카탈로그 번호수량
2284030 gels
카탈로그 번호 22840
제품 가격(KRW)
236,000
온라인 행사
Ends: 31-Dec-2026
269,000
할인액 33,000
Each
수량:
30 gels
대량 주문 또는 맞춤형 요청
제품 가격(KRW)
236,000
온라인 행사
Ends: 31-Dec-2026
269,000
할인액 33,000
Each

The Thermo Scientific™ Pierce Power Staining kits are designed for fast Coomassie dye staining of protein gels. The Pierce Mini Gel Power Staining Kit enables rapid Coomassie dye staining of proteins mini-size polyacrylamide gels and the removal of unbound stain from the gel matrix. Two mini gels having the same formulation can be accommodated and stained simultaneous in the same power stain cassette.

  • High performance—performance is equivalent to, or better than, traditional Coomassie dye staining techniques
  • Fast—optimized ionic solutions enable rapid Coomassie dye staining of protein and gel destaining.
  • Compatible—verified to work with commonly used precast and homemade SDS-PAGE gels

Traditional Coomassie dye staining techniques require staining for one hour to overnight and prolonged destaining for desired results. When used together, the Pierce Power stain solution, destain solution, and gel pads enable efficient staining and destaining in minutes with results equivalent to, or better than, traditional Coomassie dye staining techniques.

For Research Use Only. Not for use in diagnostic procedures.

사양
설명Mini Gel Power Staining Kit, 30 Gels, Coomassie Dye, Protein Gel Electrophoresis Application, Colorimetric Detection Method, In-Gel Detection, The Kit Contains Sufficient Reagents To Stain 30 Mini-Sized Gels at 15mL Each
검출 위치In-Gel Detection
검출 방법Colorimetric
제제Proprietary gel stain and destain solutions
수량30 gels
충분30 mini-sized gels at 15mL each
타겟 분자Protein
라벨 또는 염료Coomassie
제품라인Pierce
제품 유형Fast Protein Gel Stain
Unit SizeEach
구성 및 보관
Note: The kit contains sufficient reagents to stain 30 mini-sized gels at 15 mL each.

Contents:
Mini Staining Gel Pads, 2 boxes of 30 each (8 sheets/pad)
Pierce Power Stain Solution, 480 mL
Destain Solution, 480 mL

자주 묻는 질문(FAQ)

What is an alternative product to the Pierce Mini Gel Power Staining Kit (Cat. No 22840)?

No, we do not have an alternative product to the Pierce Mini Gel Power Staining Kit (Cat. No 22840). There is just the Pierce Midi Gel Power Staining Kit (Cat. No. 22839), which is not an alternative but just a different gel size.

Find additional tips, troubleshooting help, and resources within our Protein Gel Electrophoresis Chambers, Power Supplies, and Accessories Support Center.