Podophyllotoxin, 95%
Podophyllotoxin, 95%
Podophyllotoxin, 95%
Acros Organics

Podophyllotoxin, 95%

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
1 g
Catalog number 228411000
also known as 22841
HSN 29322090
Price (INR)
11,727.00
Each
Quantity:
100 mg
Request bulk or custom format
Price (INR)
11,727.00
Each
Chemical Identifiers
CAS518-28-5
IUPAC Name(10R,11R,15R,16R)-16-hydroxy-10-(3,4,5-trimethoxyphenyl)-4,6,13-trioxatetracyclo[7.7.0.0³,⁷.0¹¹,¹⁵]hexadeca-1,3(7),8-trien-12-one
Molecular FormulaC22H22O8
InChI KeyYJGVMLPVUAXIQN-XVVDYKMHSA-N
SMILESCOC1=CC(=CC(OC)=C1OC)[C@H]1[C@@H]2[C@H](COC2=O)[C@@H](O)C2=CC3=C(OCO3)C=C12
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=94 %
Appearance (Color)White to off-white
Appearance (Form)Powder
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only