Mini Gel Incubation Trays

Catalog number: 22843

Thermo Scientific™  Related applications: Protein Assays and Analysis | Protein Gel Electrophoresis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Mini Gel Incubation Trays are a convenient way to process mini-sized protein gels and Western blot membranes for use in general immunostaining applications. The trays are reusable, optically clear, and supplied with a sealable lid.

Dimensions:
• Height: 2.5 cm
• Reservoir (at the base): 10.1 cm x 9.5 cm
• Container (incl. the rim): 13.4 cm x 12.4 cm
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): Pierce™ Power System
Product Size: 10 each
Type: Tray

Documents

Manuals & protocols

Recommended products