Hidrobromuro de 3,4-dihidroxibencilamina, 98 %, Thermo Scientific Chemicals
Hidrobromuro de 3,4-dihidroxibencilamina, 98 %, Thermo Scientific Chemicals
Hidrobromuro de 3,4-dihidroxibencilamina, 98 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Hidrobromuro de 3,4-dihidroxibencilamina, 98 %, Thermo Scientific Chemicals

CAS: 16290-26-9 | C7H10BrNO2 | 220.07 g/mol
Have Questions?
Número de catálogoCantidadEnvase
2285700505 gFrasco de vidrio
Número de catálogo 228570050
Precio (USD)
-
Cantidad:
5 g
Envase:
Frasco de vidrio
Pedido a granel o personalizado
Identificadores químicos
CAS16290-26-9
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Color)Light beige
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Titration with HClO4>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only