L(-)-Histidinol dihydrochloride, 99%
L(-)-Histidinol dihydrochloride, 99%
L(-)-Histidinol dihydrochloride, 99%
Thermo Scientific Chemicals

L(-)-Histidinol dihydrochloride, 99%

CAS: 1596-64-1 | C6H13Cl2N3O | 214.09 g/mol
Have Questions?
Catalog NumberQuantity
2288300101 g
Catalog number 228830010
Price (USD)
160.65
Special offer
Online exclusive
Ends: 28-Dec-2026
189.00
Save 28.35
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
160.65
Special offer
Online exclusive
Ends: 28-Dec-2026
189.00
Save 28.35
Each
Chemical Identifiers
CAS1596-64-1
IUPAC Namedihydrogen (2S)-2-amino-3-(1H-imidazol-5-yl)propan-1-ol dichloride
Molecular FormulaC6H13Cl2N3O
InChI KeyFRCAFNBBXRWXQA-XRIGFGBMSA-N
SMILES[H+].[H+].[Cl-].[Cl-].N[C@H](CO)CC1=CN=CN1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Melting point194°C to 200°C
Titration Argentometric>=98.5 %
Appearance (Color)White to light beige
Appearance (Form)Crystalline powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only