Creatinine, 98%
Creatinine, 98%
Creatinine, 98%
Thermo Scientific Chemicals

Creatinine, 98%

CAS: 60-27-5 | C4H7N3O | 113.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
50 g
250 g
Packaging:
Glass bottle
Catalog number 228940500
Price (USD)
104.00
Each
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
104.00
Each
Chemical Identifiers
CAS60-27-5
IUPAC Name2-amino-1-methyl-4,5-dihydro-1H-imidazol-4-one
Molecular FormulaC4H7N3O
InChI KeyDDRJAANPRJIHGJ-UHFFFAOYSA-N
SMILESCN1CC(=O)N=C1N
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Loss on drying=<3.0 % (105°C, 3 h)
Appearance (Form)Crystalline powder
HPLC>=97.5 %
Sulfated ash=<0.2 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only