3-Hydroxybenzylhydrazine dihydrochloride, 98%, Thermo Scientific Chemicals
3-Hydroxybenzylhydrazine dihydrochloride, 98%, Thermo Scientific Chemicals
3-Hydroxybenzylhydrazine dihydrochloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Hydroxybenzylhydrazine dihydrochloride, 98%, Thermo Scientific Chemicals

CAS: 81012-99-9 | Molecular Formula: C7H12Cl2N2O | Molecular Weight: 211.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 229030010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS81012-99-9
IUPAC Namedihydrogen 3-(hydrazinylmethyl)phenol dichloride
Molecular FormulaC7H12Cl2N2O
InChI KeyONOJPUDFIOEGCX-UHFFFAOYSA-N
SMILES[H+].[H+].[Cl-].[Cl-].NNCC1=CC=CC(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to beige
Appearance (Form)Adhering powder
Infrared spectrumConforms
Titration Argentometric>=97.5 %

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only