Brilliant Green, pure, high purity biological stain
Brilliant Green, pure, high purity biological stain
Brilliant Green, pure, high purity biological stain
Thermo Scientific Chemicals

Brilliant Green, pure, high purity biological stain

CAS: 633-03-4 | C27H34N2O4S | 482.639 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number 229605000
Price (USD)
299.00
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
299.00
Each
Chemical Identifiers
CAS633-03-4
IUPAC Namebis[4-(diethylamino)phenyl](phenyl)methylium hydrogen sulfate
Molecular FormulaC27H34N2O4S
InChI KeyNNBFNNNWANBMTI-UHFFFAOYSA-M
SMILESOS([O-])(=O)=O.CCN(CC)C1=CC=C(C=C1)[C+](C1=CC=CC=C1)C1=CC=C(C=C1)N(CC)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Green to green-yellow
UV Lambda max625 to 631 nm (in 50% ethanol)
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
Loss on drying=<5 % (105°C)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only