Thermo Scientific™
LifterSlips™
 Related applications:
Anatomical Pathology
Achieve hybridization uniformity with Thermo Scientific™ LifterSlips™, made of premium printed cover glass. Allows for even dispersal of hybridization solutions between the substrate and cover slip.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
Features:
  • Raised edge design provides separation
  • Printed bars allow easy injection of hybridization solutions under prepositioned cover slips
  • Capillary action ensures slips stay in place
Ordering Information:
Packed per 3 doz. in boxes; minimum order quantity. Other dimensions available on request.
Documents
Manuals & protocols