mSeries LifterSlips™
 Related applications:
Anatomical Pathology
Solve the problem of hybridization nonuniformity with Thermo Scientific™ mSeries LifterSlips™. These slips are manufactured from 1mm glass, specifically drawn for flatness and superior surface quality.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
Features:
  • Precise ink printing provides innovative raised edge design for consistent dispersal and easy injection of hybridization solution under the prepositioned LifterSlip
  • Increased stiffness reduces variation in hybridization fluid volume
  • Substrate does not flex or bow
Ordering Information:
  • Packed per 50 pieces in boxes
  • Three boxes minimum order
  • Other dimensions available on request
Documents
Manuals & protocols