AimTab™ Reducing Substances Tablets
Germaine™ Laboratories

AimTab™ Reducing Substances Tablets

For the semi-quantitative determination of the amount of reducing substances (glucose, lactose, fructose, galactose, pentoses) in urine, stool, wine, or any other liquids. Tablets provides clinically useful information on carbohydrate metabolism. FOR FORENSIC USE ONLY.
Have Questions?
Número de catálogoCantidad
23-111-35836 comprimidos
Número de catálogo 23-111-358
Precio (USD)
-
Cantidad:
36 comprimidos
For the semi-quantitative determination of the amount of reducing substances (glucose, lactose, fructose, galactose, pentoses) in urine, stool, wine, or any other liquids. Tablets provides clinically useful information on carbohydrate metabolism. FOR FORENSIC USE ONLY.
  • Color chart included for easy to read results
  • Can be used to perfom the 5-Drop Method or 2-Drop Method
  • Store Between 15° to 30°C
  • Do not store in sunlight or humid places
Especificaciones
Para utilizar con (aplicación)For urine, stool or other liquids.
Tipo de empaquetadoBottle
Cantidad36 comprimidos
Duración de almacenamiento12 to 16 Months
Marcado CENo
Analitos detectablesGlucose, Lactose, Fructose, Galactose, Pentose
Unit SizeEach
Contenido y almacenamiento
De 15 °C a 30 °C