PROTOCOL™ Parasitology Three-Vial Kits, SAF/Clean/C and S
Actual product may vary
Thermo Scientific™

PROTOCOL™ Parasitology Three-Vial Kits, SAF/Clean/C and S

Standardize procedures for the routine collection, safe transportation, preservation and examination of stool specimens for intestinal parasites and enteric pathogens.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberMedia Type
2300532Zn-PVA, Formalin, C&S
2300530Modified (Cu) PVA, Formalin, Clean
2300526SAF, C&S, Clean
2300528Zn-PVA, Formalin, Clean
Catalog number 2300532
Price (CLP)
-
Media Type:
Zn-PVA, Formalin, C&S
Standardize procedures for the routine collection, safe transportation, preservation and examination of stool specimens for intestinal parasites and enteric pathogens with Thermo Scientific™ PROTOCOL™ Parasitology Three-Vial Kits.

Zinc Sulfite-Polyvinyl Alcohol (Zn-PVA):

  • Mercury-free fixative is for use as a preservative/fixative for protozoan trophozoites
  • Slides fixed with Zn-PVA may be stained using trichrome or hematoxylin

Sodium Acetate Acetic Acid Formalin (SAF):

  • Mercury-free preservative/fixative is ideally suited for use with helminth eggs and larvae, protozoan trophozoites and cysts, coccidian oocysts and microsporidian spores
  • Slides fixed with SAF may be stained using trichrome or hematoxylin

10% Buffered Formalin:

  • All-purpose fixative is appropriate for helminth eggs and larvae, and for protozoan cysts
  • Buffering system helps maintain organism morphology
  • Fixed specimens may be examined directly or concentrated for recovery of eggs, larvae and protozoan cysts

Modified Cupric Sulfate-Polyvinyl Alcohol (Modified Cu-PVA):

  • Fixative acts as a preservative/fixative for protozoan trophozoites
  • Slides fixed with Modified PVA may be stained using trichrome or hematoxylin

Culture and Sensitivity (C & S Media):

  • C&S is for collecting and transporting enteric bacterial pathogens such as Yersinia, Salmonella, Shigella, Campylobacter, and CholeraVibrio
  • Bacterial population is maintained for optimum recovery, up to 96 hours

Clean Vials:

  • Empty 30mL vials are for transporting specimens

Ordering Alerts:

Supplied as 10 packs of three vials each

Note:

All PROTOCOL™ Parasitology products are mercury free.

For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Media TypeZn-PVA, Formalin, C&S
Quantity10 Kits/Pk.
Volume (English)1.01 oz.
Volume (Metric)30 mL
Product TypeParasitology Three Vial Kit
Unit SizeEach