PROTOCOL™ Parasitology Single-Vial Kits, Clean Vials
Thermo Scientific™

PROTOCOL™ Parasitology Single-Vial Kits, Clean Vials

Standardize procedures for the routine collection, safe transportation, preservation and examination of stool specimens for intestinal parasites and enteric pathogens.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberMedia Type
2300543C&S
230054610% Buffered Formalin
2300533Modified (Cu) PVA
2300541SAF
2300537Zn-PVA
2300531Clean
2300547Cary-Blair
Catalog number 2300543
Price (CLP)
-
Media Type:
C&S
Standardize procedures for the routine collection, safe transportation, preservation and examination of stool specimens for intestinal parasites and enteric pathogens with Thermo Scientific™ PROTOCOL™ Parasitology Single-Vial Kits.

Zinc Sulfite-Polyvinyl Alcohol (Zn-PVA):

  • Mercury-free fixative for can be used as a preservative/fixative for protozoan trophozoites
  • Slides fixed with Zn-PVA can be stained using trichrome or hematoxylin

Sodium Acetate Acetic Acid Formalin (SAF):

  • Mercury-free preservative/fixative are ideally suited for use with helminth eggs and larvae, protozoan trophozoites and cysts, coccidian oocysts and microsporidian spores
  • Slides fixed with SAF can be stained using trichrome or hematoxylin

10% Neutral Buffered Formalin:

  • All-purpose fixative is appropriate for helminth eggs and larvae, and for protozoan cysts
  • Buffering system helps maintain organism morphology
  • Fixed specimens can be examined directly or concentrated for recovery of eggs, larvae and protozoan cysts

Modified Cupric Sulfate-Polyvinyl Alcohol (Modified Cu-PVA):

  • Preservative acts as a fixative for protozoan trophozoites
  • Slides fixed with Modified PVA may be stained using trichrome or hematoxylin

Culture and Sensitivity (C & S Media):

  • C&S can be used for collecting and transporting enteric bacterial pathogens such as Yersinia, Salmonella, Shigella, Campylobacter and Cholera Vibrio
  • Bacterial population is maintained for optimum recovery, up to 96 hours

Cary-Blair Media:

  • Non-nutritive medium maintains the bacterial population of enteric pathogens such as Yersinia, Salmonella, Shigella, Campylobacter and Cholera Vibrio for up to 96 hours

Clean Vials:

  • Empty 30mL vials are for transporting specimens

Ordering Alerts:

Supplied as pack of 20 vials

Note:

All PROTOCOL Parasitology products are mercury free.

For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Media TypeC&S
Quantity1 Vial/Kit, 20 Vials/Pk.
Volume (English)1.01 oz.
Volume (Metric)30 mL
Product TypeParasitology Single Vial Kit
Unit SizeEach