D(+)-Biotin, 98%
D(+)-Biotin, 98%
D(+)-Biotin, 98%
Thermo Scientific Chemicals

D(+)-Biotin, 98%

CAS: 58-85-5 | C10H16N2O3S | 244.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
1 g
5 g
25 g
Packaging:
View all
Glass Bottle
Catalog number 230090010
Price (EUR)
120,65
Online exclusive
134,00
Save 13,35
Each
Quantity:
1 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (EUR)
120,65
Online exclusive
134,00
Save 13,35
Each
Chemical Identifiers
CAS58-85-5
IUPAC Name5-{2-oxo-hexahydro-1H-thieno[3,4-d]imidazol-4-yl}pentanoic acid
Molecular FormulaC10H16N2O3S
InChI KeyYBJHBAHKTGYVGT-UHFFFAOYNA-N
SMILESOC(=O)CCCCC1SCC2NC(=O)NC12
View more
SpecificationsSpecification SheetSpecification Sheet
Specific optical rotation+89° to +93° (20°C, 589 nm) (c=2, 0.1 N NaOH)
Specific optical rotationon dried substance
Appearance of solution(2% soln. in 0.1 N NaOH) clear, colorless
Appearance (Color)White to off-white
Appearance (Form)Powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only