Laminin Mouse Protein, Natural - FAQs
Laminin Mouse Protein, Natural - FAQs
View additional product information for Laminin Mouse Protein, Natural - FAQs (23017015)
5 product FAQs found
What is the molecular weight of Laminin Mouse Protein, Natural?
It has 3 subunits and has a MW of 850 kDa: one A chain is 400 kDa and two B chains are 205 and 215 kDa.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
I would like to know how Laminin Mouse Protein, Natural was sterilized.
Laminin Mouse Protein, Natural (Cat. No. 23017015) is sterile filtered through a 0.22 micron filter membrane.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Can I use the Poly-D-Lysine matrix with other coatings?
iPSC-derived neurons have been cultured successfully on Poly-D-Lysine with a secondary coating of mouse and human laminin.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Can natural mouse laminin be thawed, aliquoted, and refrozen?
Yes, it is recommended to store usage size aliquots at -68 degrees C to -85 degrees C.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What is the protein concentration per vial for natural mouse laminin?
The protein concentration per vial is 1 mg/vial. The specification for protein concentration can range from 0.5 to 2.0 mg/mL. The volume per vial can vary as it will be based on the protein concentration determined for the specific lot. The volume added to each vial will equal 1 mg of laminin.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.