Bilirubin, 98+%
Bilirubin, 98+%
Bilirubin, 98+%
Thermo Scientific Chemicals

Bilirubin, 98+%

Bilirubin, CAS # 635-65-4, is a tetrapyrroliccompound found in bile that is a product of heme catabolism. | CAS: 635-65-4 | C33H36N4O6 | 584.67 g/mol
数量:
1 g
5 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 230220050
価格(JPY)
-
見積もりを依頼する
数量:
5 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS635-65-4
IUPAC Name3-(2-{[3-(2-carboxyethyl)-5-{[(2Z)-4-ethenyl-3-methyl-5-oxo-2,5-dihydro-1H-pyrrol-2-ylidene]methyl}-4-methyl-1H-pyrrol-2-yl]methyl}-5-{[(2Z)-3-ethenyl-4-methyl-5-oxo-2,5-dihydro-1H-pyrrol-2-ylidene]methyl}-4-methyl-1H-pyrrol-3-yl)propanoic acid
Molecular FormulaC33H36N4O6
InChI KeyBPYKTIZUTYGOLE-IFADSCNNSA-N
SMILESCC1=C(C=C)\C(NC1=O)=C\C1=C(C)C(CCC(O)=O)=C(CC2=C(CCC(O)=O)C(C)=C(N2)\C=C2/NC(=O)C(C=C)=C2C)N1
さらに表示
UV spectrophotometry>=98.0 %
Molar absorption coefficient>=58800 l/(mol.cm) (at Lambda max, in CHCl3)
Appearance (Color)Orange to red to brown
Infrared spectrumConforms
UV Lambda max450 to 454 nm (in CHCl3)
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Bilirubin is a degradation product of heme following the activity of heme oxidase. Bilirubin is produced in the reticuloendothelial system and conjugated in the liver. It appears as an orange-yellow pigment that is secreted in the bile
  • High levels of unconjugated bilirubin can result in jaundice, a condition of the yellowing of skin and eyes. This can be toxic and may be a sign of severe liver disease
  • Bilirubin has been seen to have functions including cell signaling, metabolism modulation, and immune regulation
  • It has protective antioxidant activity at moderate levels

Application

  • Bilirubin is widely used in studies of liver function and disease, such as hepatitis or cirrhosis
  • As an antioxidant, bilirubin has been studied in protection against H2O2-mediated cell death
  • It can be used as a scavenger of reactive oxygen species (ROS) or as an inhibitor of NADPH oxidase
RUO – Research Use Only

General References:

  1. Hinds TD Jr, Stec DE. Bilirubin, a Cardiometabolic Signaling Molecule. Hypertension. 2018 Oct;72(4):788-795. doi: 10.1161/HYPERTENSIONAHA.118.11130. Erratum in: Hypertension. 2018 Dec;72(6):e95. PMID: 30354722; PMCID: PMC6205727.
  2. Vítek L, Tiribelli C. Bilirubin: The yellow hormone? J Hepatol. 2021 Dec;75(6):1485-1490. doi: 10.1016/j.jhep.2021.06.010. Epub 2021 Jun 18. PMID: 34153399.