L(-)-Carnitine hydrochloride, 98%
L(-)-Carnitine hydrochloride, 98%
L(-)-Carnitine hydrochloride, 98%
Thermo Scientific Chemicals

L(-)-Carnitine hydrochloride, 98%

CAS: 6645-46-1 | C7H16ClNO3 | 197.66 g/mol
製品番号(カタログ番号)数量
23028025025 g
製品番号(カタログ番号) 230280250
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS6645-46-1
IUPAC Name[(2R)-3-carboxy-2-hydroxypropyl]trimethylazanium chloride
Molecular FormulaC7H16ClNO3
InChI KeyJXXCENBLGFBQJM-FYZOBXCZSA-N
SMILES[Cl-].C[N+](C)(C)C[C@H](O)CC(O)=O
さらに表示
Loss on drying=<5 %
Appearance (Color)White to beige
Specific optical rotation-21° to -24° (20°C, 589 nm) (c=1, H2O)
Appearance (Form)Crystalline powder
Infrared spectrumConforms
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only