Pierce™ NEM (N-ethylmaleimide)

Catalog number: 23030B

Thermo Scientific™  Related applications: Protein Labeling & Crosslinking

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Thermo Scientific Pierce N-Ethylmaleimide (NEM) is a small compound that forms stable, covalent thioether bonds with sulfhydryls (e.g., reduced cysteines), enabling them to be permanently blocked to prevent disulfide bond formation.

Features of N-Ethylmaleimide (NEM):

• Permanently block sulfhydryls to prevent disulfide bond formation.
• Monitor the reaction by measuring the decrease in absorbance at 300nm
• Block sulfhydryl-containing reagents that interfere in enzyme assays

N-Ethylmaleimide (NEM) is an alkylating reagent that reacts with sulfhydryls. At pH 6.5-7.5, the maleimide reaction is specific for sulfhydryls; however, at pH values greater than 7.5, reactivity with amino groups occurs. Maleimide groups react with sulfhydryls by nucleophilic attack of the thiolate anion on one of the carbons of the double bond. When sufficient sulfhydryls have been blocked, the reaction can be monitored by measuring the decrease in absorbance at 300nm as the double bond becomes a single bond. The resulting thioether group is non-reversible and terminates in an ethyl group, blocking or capping the sulfhydryl.

Related Products
Pierce™ MMTS (methyl methanethiosulfonate)
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Chemical Reactivity: Thiol
Label or Dye: Unreactive Group
Product Line: Pierce™
Product Size: Custom
Solubility: Water
Spacer: Mid-Length

Contents & storage

Upon receipt store at -20°C.

Documents

Manuals & protocols