Folinic acid, calcium salt pentahydrate, 95.0-105.0%
Folinic acid, calcium salt pentahydrate, 95.0-105.0%
Folinic acid, calcium salt pentahydrate, 95.0-105.0%
Thermo Scientific Chemicals

Folinic acid, calcium salt pentahydrate, 95.0-105.0%

CAS: 6035-45-6 | C20H21CaN7O7·5H2O | 601.58 g/mol
数量:
250 mg
1 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 230312500
価格(JPY)
-
数量:
250 mg
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS6035-45-6
IUPAC Name(2S)-2-[(4-{[(2-amino-5-formyl-4-oxo-1,4,5,6,7,8-hexahydropteridin-6-yl)methyl]amino}phenyl)formamido]pentanedioic acid calcium pentahydrate
Molecular FormulaC20H33CaN7O12
InChI KeyDZNHRKNLTYYMQA-ZIGBGYJWSA-N
SMILESO.O.O.O.O.[Ca].NC1=NC(=O)C2=C(NCC(CNC3=CC=C(C=C3)C(=O)N[C@@H](CCC(O)=O)C(O)=O)N2C=O)N1
さらに表示
Appearance (Form)Powder
Appearance (Color)White to yellow
Infrared spectrumConforms
HPLC>=95 %
Water=<17 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only