1-O-n-Octyl-beta-D-glucopyranoside, 98%
1-O-n-Octyl-beta-D-glucopyranoside, 98%
1-O-n-Octyl-beta-D-glucopyranoside, 98%
Thermo Scientific Chemicals

1-O-n-Octyl-beta-D-glucopyranoside, 98%

CAS: 29836-26-8 | C14H28O6 | 292.37 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 230340250
Price (EUR)
1.137,65
Online exclusive
1.264,00
Save 126,35 (10%)
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
1.137,65
Online exclusive
1.264,00
Save 126,35 (10%)
Each
Chemical Identifiers
CAS29836-26-8
IUPAC Name(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-(octyloxy)oxane-3,4,5-triol
Molecular FormulaC14H28O6
InChI KeyHEGSGKPQLMEBJL-RKQHYHRCSA-N
SMILESCCCCCCCCO[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Infrared spectrumConforms
Solubility(10 % in water) Clear to slightly hazy colorless to light yellow
HPLC>=97.5 %
Appearance (Form)Powder and/or chunks
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only