This product ships via ground from US northeast.
Dodecyl sulfate, sodium salt, 99%, for biochemistry
Dodecyl sulfate, sodium salt, 99%, for biochemistry
Dodecyl sulfate, sodium salt, 99%, for biochemistry
Thermo Scientific Chemicals

Dodecyl sulfate, sodium salt, 99%, for biochemistry

CAS: 151-21-3 | C12H25NaO4S, C12H25NaO4S | 288.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
100 g
500 g
2.5 kg
Catalog number 230425000
Price (USD)
254.65
Special offer
Online exclusive
Ends: 28-Dec-2026
299.00
Save 44.35
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
254.65
Special offer
Online exclusive
Ends: 28-Dec-2026
299.00
Save 44.35
Each
Chemical Identifiers
CAS151-21-3
IUPAC Namesodium dodecyl sulfate
Molecular FormulaC12H25NaO4S
InChI KeyDBMJMQXJHONAFJ-UHFFFAOYSA-M
SMILES[Na+].CCCCCCCCCCCCOS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Loss on drying=<1 % (1 g, 105°C, 3 h)
UV(3 % in water)
Solubility(3 % in water) Clear colorless
Chloride (Cl)=<500 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For biochemistry
RUO – Research Use Only