StemPro™ EZPassage™ Disposable Stem Cell Passaging Tool
Gibco™
StemPro™ EZPassage™ Disposable Stem Cell Passaging Tool
The StemPro™ EZPassage™ Disposable Stem Cell Passaging Tool is a novel human embryonic stem cell passaging device that makes manualpassagingRead more
Have Questions?
Catalog NumberQuantity
2318101010 units
Catalog number 23181010
Price (KRW)
441,000
Online offer
Ends: 15-Jul-2026
489,000
Save 48,000 (10%)
Each
Quantity:
10 units
Price (KRW)
441,000
Online offer
Ends: 15-Jul-2026
489,000
Save 48,000 (10%)
Each
The StemPro™ EZPassage™ Disposable Stem Cell Passaging Tool is a novel human embryonic stem cell passaging device that makes manual
passaging of stem cell colonies more rapid and reproducible. The Passaging Tool is made of a proprietary, cell culture-safe, and inert material that enables cutting of embryonic stem cell colonies growing on feeder cells or other substrates into uniform pieces for optimal passaging. This tool outperforms existing manual and enzymatic passing methods in speed, uniformity of passaged colonies, and reliability.
Application:
By enabling the manual passaging of human embryonic stem cells, the StemPro EZPassage Disposable Stem Cell Passaging Tool provides
the following advantages:
–Manually passage stem cell colonies in a fraction of the time compared with standard techniques.
–Cut stem cell colonies into pieces of uniform size, making passaging of stem cells more reproducible.
–Control the size for better passaging or to make embryoid bodies.
–Cell culture-safe Passaging Tools are ready for use and packaged individually in gamma-irradiated sleeves.
The StemPro EZPassage Disposable Stem Cell Passaging Tool is intended for one-time use. Do not sterilize (alcohol or autoclave) the Passaging Tool, since it may lose shape and cease to function properly.
Specifications
Cell TypeStem Cell
For Use With (Application)Manual Passaging Stem Cell Colonies
Product LineEZPassage, StemPro
Product TypeStem Cell Passaging Tool
Quantity10 units
SterilitySterile
DisposableSingle-use
MaterialProprietary, Cell Culture-Safe, Inert Material
Unit SizeEach
Frequently asked questions (FAQs)
In the StemPro EZPassage Disposable Stem Cell Passaging Tool (Cat. No. 23181010), what is the exact distance between the blades of the roller?
The tool has ˜230 micrometers between each blade. We don't recommend using the tool on anything smaller than a 6-well plate. As far as the thickness at the base of each blade relative to the distance between the blade's edges, we don't have data on that.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.