1-Propanol, 99.5%, for analysis
1-Propanol, 99.5%, for analysis
1-Propanol, 99.5%, for analysis
Thermo Scientific Chemicals

1-Propanol, 99.5%, for analysis

CAS: 71-23-8 | C3H8O | 60.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
10 L
Packaging:
View all
Glass Bottle
Catalog number 232070025
Price (EUR)
127,65
Online Exclusive
142,00
Save 14,35 (10%)
Each
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (EUR)
127,65
Online Exclusive
142,00
Save 14,35 (10%)
Each
Chemical Identifiers
CAS71-23-8
IUPAC Namepropan-1-ol
Molecular FormulaC3H8O
InChI KeyBDERNNFJNOPAEC-UHFFFAOYSA-N
SMILESCCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<10 APHA
2-Propanol=<0.05 % (GC)
Aluminium (Al)=<0.5 ppm
Calcium (Ca)=<0.5 ppm
Magnesium (Mg)=<0.1 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
RUO – Research Use Only