Pierce™ Bovine Serum Albumin Standard, 2 mg/mL

Catalog number: 23210B

Thermo Scientific™  Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Thermo Scientific Pierce BSA Protein Assay Standards are high-quality reference samples for generating accurate standard curves and calibration controls in total protein assays.

Features of BSA Protein Assay Standards:

Universal—recognized as the industry standard for protein quantitation in colorimetric protein assays
Pure and stable—supplied in ultrapure 0.9% saline solution with 0.05% sodium azide; room temperature stable
Accurate and consistent—precisely formulated at 2.00 +/-0.03 mg/mL compared to an NIST reference

These bovine serum albumin (BSA) solutions are protein concentration reference standards for use in BCA, Bradford and other protein assay protocols. BSA is the universally accepted reference protein for total protein quantitation. The albumin standard is precisely formulated at 2 mg/mL in an ultrapure 0.9% sodium chloride (saline) solution. The concentration of the stock solution is calibrated by direct comparison to purified BSA (Fraction V) from the National Institute of Standards and Technology (NIST).

Applications:
• Protein assay quantitation standard (BCA Protein Assay, Coomassie-Bradford Assay, etc.)
• Protein recovery control for desalting and other column procedures
• General calibration of spectrophotometer UV-lamp (absorbance at 280nm)

Selection of a protein standard is potentially the greatest source of error in any protein assay. The best choice for a standard is a purified, known concentration of the most abundant protein contained in the samples being tested. Often, a highly purified, known concentration of the protein of interest is not available or it is too expensive to use as the standard, or the sample itself is a mixture of many proteins (e.g., cell lysate). In such cases, the best standard is one that will produce a normal (i.e., average) color response curve with the selected protein assay method and is readily available to any researcher. BSA is such a protein, and the Pierce Albumin Standards are the most convenient source of ready-to-use BSA standard.

For greatest accuracy in estimating total protein concentration in unknown samples, it is essential to include a standard curve each time the assay is performed. This is particularly true for the protein assay methods that produce non-linear standard curves. Deciding on the number of standards and replicates used to define the standard curve depends upon the degree of non-linearity in the standard curve and the degree of accuracy required. In general, fewer points are needed to construct a standard curve if the color response is linear. Typically, standard curves are constructed using at least two replicates for each point on the curve.

Related Products
Pierce™ Bovine Serum Albumin Standard Pre-Diluted Set
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Assay Type: Protein Quantitation Assay
Product Line: Pierce™
Product Size: Custom
Technique: Internal Standard

Contents & storage

Store at 2–8°C.

Documents

Manuals & protocols